Assay Details
| Gene Symbol: Spag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012116.1 | NP_001012116.1 | 17 - 18 | 2490 | 97 | |
| XM_008765453.1 | XP_008763675.1 | 17 - 18 | 2710 | 97 | ||
| GenBank mRNA | BC085828.1 | - | 17 - 18 | 2490 | 7132502 | 97 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P104488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80895 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P104488 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P104488 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P104488 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P187732 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73007 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92968 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1512 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9299 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10896195 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896183 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896182 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896197 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896190 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896186 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896185 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896188 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896196 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896200 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896199 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896192 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896193 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896198 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896184 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896194 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896191 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896189 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896187 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10896181 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700007389.rn.1 | TC0700000970.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700007390.rn.1 | TC0700000970.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700007388.rn.1 | TC0700000970.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10896181 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21401792 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376354 | - | - |

Spag1
Genomic Map