Assay Details
| Gene Symbol: Grina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153308.4 | NP_695220.4 | 1 - 2 | 168 | 96 | |
| GenBank mRNA | BC062074.1 | - | 1 - 2 | 138 | 5599428 | 96 |
| CK475470.1 | - | 1 - 2 | 168 | 96 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC062074.1,NM_153308.4 transcripts.
Set Membership: |
|

Grina
Genomic Map