Assay Details
| Gene Symbol: Grina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153308.4 | NP_695220.4 | 4 - 5 | 814 | 66 | |
| GenBank mRNA | BC062074.1 | - | 4 - 5 | 784 | 5599428 | 66 |
More Information
Updated Mapping Information
Set Membership: |
|

Grina
Genomic Map