Assay Details
| Gene Symbol: Cenpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130504.1 | NP_001123976.1 | 2 - 3 | 201 | 65 | |
| XM_006242086.2 | XP_006242148.1 | 2 - 3 | 466 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cenpm
Genomic Map