Assay Details
| Gene Symbol: Cenpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130504.1 | NP_001123976.1 | 3 - 4 | 297 | 128 | |
| XM_006242086.2 | XP_006242148.1 | 3 - 4 | 562 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cenpm
Genomic Map