Assay Details
| Gene Symbol: Cenpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130504.1 | NP_001123976.1 | 4 - 5 | 377 | 96 | |
| XM_008765720.1 | XP_008763942.1 | 3 - 4 | 539 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cenpm
Genomic Map