Assay Details
| Gene Symbol: LOC100911597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750378.3 | XP_003750426.1 | 1 - 2 | 215 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003750378.3 transcripts.
Set Membership: |
|

LOC100911597
Genomic Map