Assay Details
| Gene Symbol: LOC100911597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750378.3 | XP_003750426.1 | 21 - 22 | 2863 | 65 | |
| Gene Symbol: Myh9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013194.1 | NP_037326.1 | 2648 | 65 | ||
| GenBank mRNA | U31463.1 | - | 2648 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911597
Genomic Map