Assay Details
| Gene Symbol: Lynx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130546.1 | NP_001124018.1 | 1 - 2 | 127 | 157 | |
| GenBank mRNA | FQ212028.1 | - | 1 - 2 | 135 | 157 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001130546.1 transcripts.
Set Membership: |
|

Lynx1
Genomic Map