Assay Details
| Gene Symbol: Enthd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135913.1 | NP_001129385.1 | 3 - 4 | 791 | 84 | |
| XM_006242122.2 | XP_006242184.1 | 4 - 5 | 1288 | 84 | ||
| GenBank mRNA | BC167764.1 | - | 3 - 4 | 791 | 7453894 | 84 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P156243 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P023694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41534 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P023694 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P156243 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P023694 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P156243 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P023694 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P156243 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10905555 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905556 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905557 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905553 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905552 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905554 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10905551 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700027851.rn.1 | TC0700003533.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027849.rn.1 | TC0700003533.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027844.rn.1 | TC0700003533.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027845.rn.1 | TC0700003533.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027846.rn.1 | TC0700003533.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027847.rn.1 | TC0700003533.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027853.rn.1 | TC0700003533.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10905551 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21784830 | - | - |

Enthd1
Genomic Map