Assay Details
| Gene Symbol: Zhx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271056.1 | NP_001257985.1 | 1 - 2 | 70 | 55 | |
| XM_006241654.2 | XP_006241716.1 | 1 - 2 | 705 | 55 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271056.1,XM_006241654.2 transcripts.
Set Membership: |
|

Zhx2
Genomic Map