Assay Details
| Gene Symbol: Sstr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133522.1 | NP_598206.1 | 1 - 2 | 469 | 63 | |
| GenBank mRNA | X63574.1 | - | 1 - 2 | 469 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_133522.1, X63574.1 transcripts.
Set Membership: |
|

Sstr3
Genomic Map