Assay Details
| Gene Symbol: Grap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034944.1 | NP_001030116.1 | 5 - 6 | 807 | 78 | |
| GenBank mRNA | BC105895.1 | - | 5 - 6 | 807 | 7380480 | 78 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P525607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32491 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P525607 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P525607 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P525607 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P525607 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69295 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74310 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1596 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91568 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10897806 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10897808 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10897809 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10897807 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10897811 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10897813 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10897814 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10897810 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10897812 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10897805 | - | - |
| Rat Genome 230 2.0 Array | 1391896_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391896_at | - | - |
| Affymetrix HT Rat Focus Array | 1391896_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391896_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700011163.rn.1 | TC0700001479.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700011164.rn.1 | TC0700001479.rn.1 | RTA TC |
| Rat Expression Set 230 | 1391896_at | - | - |
| Rat Expression Set 230 | 1391896_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10897805 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353462 | - | - |

Grap2
Genomic Map