Assay Details
| Gene Symbol: Baalc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144762.2 | NP_658907.1 | 2 - 3 | 619 | 114 | |
| GenBank mRNA | AB073318.1 | - | 2 - 3 | 619 | 114 | |
| AF371321.1 | - | 2 - 3 | 325 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P194803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145607 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P194803 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P194803 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P194803 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P194803 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P194806 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80511 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103420 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36267 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85623 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102302 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10896254 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896251 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896253 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896252 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896255 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10896250 | - | - |
| Rat Genome 230 2.0 Array | 1387938_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387938_at | - | - |
| Affymetrix HT Rat Focus Array | 1387938_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387938_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700007602.rn.1 | TC0700001012.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700007603.rn.1 | TC0700001012.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387938_at | - | - |
| Rat Expression Set 230 | 1387938_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10896250 | - | - |
| Rat Genome U34 Set | rc_AA850640_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21348659 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366187 | - | - |
| RatRef-12 Expression BeadChip | ILMN_52846 | - | - |

Baalc
Genomic Map