Assay Details
| Gene Symbol: March9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100601.1 | NP_001094071.1 | 2 - 3 | 574 | 64 | |
| XM_008765422.1 | XP_008763644.1 | 3 - 4 | 1549 | 64 | ||
| GenBank mRNA | AB048842.1 | - | 2 - 3 | 574 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

March9
Genomic Map