Assay Details
| Gene Symbol: Sntb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130542.1 | NP_001124014.1 | 6 - 7 | 1927 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P263015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99906 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P263015 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P263015 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P263015 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P263015 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10903824 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903817 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903820 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903818 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903821 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903823 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903819 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10903816 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700024133.rn.1 | TC0700003151.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700024131.rn.1 | TC0700003151.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10903816 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21324133 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368900 | - | - |

Sntb1
Genomic Map