Assay Details
| Gene Symbol: Commd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241924.2 | XP_006241986.1 | 1 - 2 | 386 | 73 | |
| Gene Symbol: LOC100910472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241915.2 | XP_006241977.1 | 1 - 2 | 386 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006241915.2, XM_006241924.2 transcripts.
Set Membership: |
|

Commd5
Genomic Map