Assay Details
| Gene Symbol: Zc3h7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130695.1 | NP_001124167.1 | 5 - 6 | 566 | 62 | |
| XM_006242085.2 | XP_006242147.1 | 6 - 7 | 850 | 62 | ||
| XM_008765719.1 | XP_008763941.1 | 6 - 7 | 1936 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zc3h7b
Genomic Map