Assay Details
| Gene Symbol: Wbp2nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001067725.4 | XP_001067725.2 | 4 - 5 | 430 | 62 | |
| XM_008765810.1 | XP_008764032.1 | 4 - 5 | 430 | 62 | ||
More Information
Set Membership: |
|

Wbp2nl
Genomic Map