Assay Details
| Gene Symbol: Sdr9c7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241515.2 | XP_006241577.1 | 4 - 5 | 5285 | 89 | |
| XM_006241516.2 | XP_006241578.1 | 4 - 5 | 3554 | 89 | ||
| XM_008765432.1 | XP_008763654.1 | 3 - 4 | 5657 | 89 | ||
| XM_008765433.1 | XP_008763655.1 | 4 - 5 | 4263 | 89 | ||
| GenBank mRNA | AY044435.1 | - | 1 - 2 | 256 | 89 | |
More Information
Set Membership: |
|

Sdr9c7
Genomic Map