Assay Details
| Gene Symbol: Sdr9c7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241515.2 | XP_006241577.1 | 5 - 6 | 5456 | 79 | |
| XM_006241516.2 | XP_006241578.1 | 5 - 6 | 3725 | 79 | ||
| XM_008765432.1 | XP_008763654.1 | 4 - 5 | 5828 | 79 | ||
| XM_008765433.1 | XP_008763655.1 | 5 - 6 | 4434 | 79 | ||
| GenBank mRNA | AY044435.1 | - | 2 - 3 | 427 | 79 | |
More Information
Set Membership: |
|

Sdr9c7
Genomic Map