Assay Details
| Gene Symbol: Efcab6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006242144.2 | XP_006242206.1 | 8 - 9 | 1216 | 59 | |
| XM_008765811.1 | XP_008764033.1 | 28 - 29 | 3893 | 59 | ||
| XM_008765812.1 | XP_008764034.1 | 28 - 29 | 3899 | 59 | ||
| XM_008765813.1 | XP_008764035.1 | 28 - 29 | 3915 | 59 | ||
| XM_008765814.1 | XP_008764036.1 | 28 - 29 | 3779 | 59 | ||
| XM_008765815.1 | XP_008764037.1 | 28 - 29 | 3730 | 59 | ||
More Information
Set Membership: |
|

Efcab6
Genomic Map