Assay Details
| Gene Symbol: Ly6i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139257.1 | NP_640350.1 | 1 - 2 | 74 | 127 | |
| XM_006241766.2 | XP_006241828.1 | 2 - 3 | 201 | 127 | ||
| XM_008765545.1 | XP_008763767.1 | 2 - 3 | 188 | 127 | ||
| GenBank mRNA | M30689.1 | - | 1 - 2 | 74 | 127 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the M30689.1, NM_139257.1, XM_006241766.2, XM_008765545.1 transcripts.
Set Membership: |
|

Ly6i
Genomic Map