Assay Details
| Gene Symbol: Trhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013047.3 | NP_037179.1 | 1 - 2 | 236 | 76 | |
| XM_006241611.1 | XP_006241673.1 | 1 - 2 | 267 | 76 | ||
| GenBank mRNA | X64630.1 | - | 1 - 2 | 164 | 76 | |
| X66726.1 | - | 1 - 2 | 237 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_013047.3, X64630.1, X66726.1, XM_006241611.1 transcripts.
Set Membership: |
|

Trhr
Genomic Map