Assay Details
| Gene Symbol: Mrvi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105210.1 | NP_001098680.1 | 17 - 18 | 2539 | 57 | |
| NM_001105211.1 | NP_001098681.1 | 16 - 17 | 2292 | 57 | ||
| XM_006230047.2 | XP_006230109.1 | 17 - 18 | 2537 | 57 | ||
| XM_006230048.2 | XP_006230110.1 | 18 - 19 | 2723 | 57 | ||
| XM_008759727.1 | XP_008757949.1 | 17 - 18 | 2639 | 57 | ||
| XM_008759728.1 | XP_008757950.1 | 17 - 18 | 2858 | 57 | ||
| XM_008759729.1 | XP_008757951.1 | 18 - 19 | 2320 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P092754 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P092747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862218 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167263 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P092747 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P092754 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P092754 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P092747 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P092747 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P092754 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P304509 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 87452 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1232 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64149 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10724903 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724916 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724918 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724915 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724917 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724910 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724913 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724919 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724909 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724907 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724912 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724914 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724922 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724904 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724906 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724911 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724908 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724905 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724921 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724920 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10724902 | - | - |
| Rat Genome 230 2.0 Array | 1383914_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383914_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100050314.rn.1 | TC0100006405.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100050315.rn.1 | TC0100006405.rn.1 | RTA TC |
| Rat Expression Set 230 | 1383914_PM_at | - | - |
| Rat Expression Set 230 | 1383914_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10724902 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22291470 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361633 | - | - |

Mrvi1
Genomic Map