Assay Details
| Gene Symbol: Ccdc101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114502.1 | NP_001107974.1 | 2 - 3 | 268 | 77 | |
| XM_006230231.2 | XP_006230293.1 | 2 - 3 | 374 | 77 | ||
| XM_006230232.2 | XP_006230294.1 | 2 - 3 | 278 | 77 | ||
| GenBank mRNA | FQ210727.1 | - | 2 - 3 | 243 | 77 | |
| FQ224509.1 | - | 2 - 3 | 243 | 77 | ||
| FQ232716.1 | - | 2 - 3 | 243 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc101
Genomic Map