Assay Details
| Gene Symbol: Hsd3b7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139329.1 | NP_647545.1 | 2 - 3 | 270 | 70 | |
| XM_006230212.2 | XP_006230274.1 | 2 - 3 | 656 | 70 | ||
| GenBank mRNA | AB000199.1 | - | 2 - 3 | 270 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hsd3b7
Genomic Map