Assay Details
| Gene Symbol: Maz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110319.1 | NP_001103789.1 | 4 - 5 | 1280 | 58 | |
| GenBank mRNA | EF059745.1 | - | 4 - 5 | 1280 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Maz
Genomic Map