Assay Details
| Gene Symbol: Ddias | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126294.1 | NP_001119766.1 | 1 - 2 | 72 | 81 | |
| GenBank mRNA | BC166470.1 | - | 1 - 2 | 72 | 9088340 | 81 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC166470.1, NM_001126294.1 transcripts.
Set Membership: |
|

Ddias
Genomic Map