Assay Details
| Gene Symbol: Gprc5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230109.2 | XP_006230171.1 | 3 - 4 | 1308 | 80 | |
| XM_006230110.2 | XP_006230172.1 | 3 - 4 | 1366 | 80 | ||
| XM_008759715.1 | XP_008757937.1 | 3 - 4 | 1381 | 80 | ||
| GenBank mRNA | BC166986.1 | - | 3 - 4 | 1298 | 80 | |
More Information
Set Membership: |
|

Gprc5b
Genomic Map