Assay Details
| Gene Symbol: Cd2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106297.2 | NP_001099767.1 | 3 - 4 | 402 | 54 | |
| XM_006230251.2 | XP_006230313.1 | 4 - 5 | 2164 | 54 | ||
| XM_006230252.2 | XP_006230314.1 | 4 - 5 | 2172 | 54 | ||
| XM_008759879.1 | XP_008758101.1 | 4 - 5 | 2178 | 54 | ||
| GenBank mRNA | BC168174.1 | - | 3 - 4 | 402 | 9032868 | 54 |
More Information
Updated Mapping Information
Set Membership: |
|

Cd2bp2
Genomic Map