Assay Details
| Gene Symbol: Cd2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106297.2 | NP_001099767.1 | 4 - 5 | 844 | 68 | |
| XM_006230251.2 | XP_006230313.1 | 5 - 6 | 2606 | 68 | ||
| XM_006230252.2 | XP_006230314.1 | 5 - 6 | 2614 | 68 | ||
| XM_008759879.1 | XP_008758101.1 | 5 - 6 | 2620 | 68 | ||
| GenBank mRNA | BC168174.1 | - | 4 - 5 | 844 | 9032868 | 68 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cd2bp2
Genomic Map