Assay Details
| Gene Symbol: Fam160a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005538.2 | NP_001005538.2 | 6 - 7 | 1506 | 82 | |
| XM_006229931.2 | XP_006229993.1 | 6 - 7 | 1540 | 82 | ||
| XM_006229932.2 | XP_006229994.1 | 6 - 7 | 1539 | 82 | ||
| XM_008759708.1 | XP_008757930.1 | 6 - 7 | 1540 | 82 | ||
| GenBank mRNA | BC082011.1 | - | 6 - 7 | 1492 | 7132039 | 82 |
More Information
Updated Mapping Information
Set Membership: |
|

Fam160a2
Genomic Map