Assay Details
| Gene Symbol: Apobr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109154.1 | NP_001102624.1 | 1 - 2 | 83 | 66 | |
| XM_006230319.2 | XP_006230381.1 | 1 - 2 | 196 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Apobr
Genomic Map