Assay Details
| Gene Symbol: Art2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | X99122.1 | - | 2 - 3 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the X99122.1 transcripts.
Set Membership: |
|

Art2b
Genomic Map