Assay Details
| Gene Symbol: Sept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012460.1 | NP_001012478.1 | 6 - 7 | 601 | 123 | |
| GenBank mRNA | BC089897.1 | - | 6 - 7 | 601 | 7381516 | 123 |
More Information
Updated Mapping Information
Set Membership: |
|

Sept1
Genomic Map