Assay Details
| Gene Symbol: Sytl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748924.3 | XP_003748972.1 | 18 - 19 | 3086 | 114 | |
| XM_006229687.2 | XP_006229749.1 | 18 - 19 | 6692 | 114 | ||
| XM_006229694.2 | XP_006229756.1 | 19 - 20 | 3206 | 114 | ||
| XM_006229695.2 | XP_006229757.1 | 18 - 19 | 3159 | 114 | ||
| XM_006229697.2 | XP_006229759.1 | 8 - 9 | 3280 | 114 | ||
| XM_008759651.1 | XP_008757873.1 | 19 - 20 | 6832 | 114 | ||
| XM_008759652.1 | XP_008757874.1 | 19 - 20 | 6983 | 114 | ||
| XM_008759653.1 | XP_008757875.1 | 19 - 20 | 9371 | 114 | ||
| XM_008759654.1 | XP_008757876.1 | 18 - 19 | 6784 | 114 | ||
| XM_008759655.1 | XP_008757877.1 | 17 - 18 | 6730 | 114 | ||
| XM_008759656.1 | XP_008757878.1 | 18 - 19 | 6712 | 114 | ||
| GenBank mRNA | BC168993.1 | - | 8 - 9 | 1000 | 9031555 | 114 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P21122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30021 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21122 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21122 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P21122 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P986848 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P433568 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21122 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92623 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30167 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10708602 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708612 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708609 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708611 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708613 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708607 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708596 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708606 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708594 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708592 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708595 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708605 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708593 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708608 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708603 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708604 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708610 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708597 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708599 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10708591 | - | - |
| Rat Genome 230 2.0 Array | 1390107_at | - | - |
| Rat Genome 230 2.0 Array | 1390107_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390107_at | - | - |
| Affymetrix HT Rat Focus Array | 1390107_PM_at | - | - |
| Rat Expression Set 230 | 1390107_PM_at | - | - |
| Rat Expression Set 230 | 1390107_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708591 | - | - |
| Rat Genome U34 Set | rc_AA943765_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20886623 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356423 | - | - |

Sytl2
Genomic Map