Assay Details
| Gene Symbol: LOC103694867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773816.1 | XP_008772038.1 | 10 - 11 | 1686 | 84 | |
| Gene Symbol: Gga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100519.1 | NP_001093989.1 | 11 - 12 | 1180 | 84 | |
| GenBank mRNA | BC098781.1 | - | 5 - 6 | 458 | 84 | |
| CO558907.1 | - | 575 | 84 | |||
More Information
Updated Mapping Information
Set Membership: |
|

LOC103694867
Genomic Map