Assay Details
| Gene Symbol: LOC103694867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773816.1 | XP_008772038.1 | 8 - 9 | 1463 | 65 | |
| Gene Symbol: Gga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100519.1 | NP_001093989.1 | 9 - 10 | 957 | 65 | |
| GenBank mRNA | BC098781.1 | - | 3 - 4 | 235 | 65 | |
| CO558907.1 | - | 352 | 65 | |||
More Information
Updated Mapping Information
Set Membership: |
|

LOC103694867
Genomic Map