Assay Details
| Gene Symbol: Gde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032615.2 | NP_116004.2 | 525 | 140 | ||
| GenBank mRNA | AF212861.1 | - | 470 | 140 | ||
| BC070897.1 | - | 525 | 7099596 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gde1
Genomic Map