Assay Details
Gene Symbol: LOC691909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134694.2 | NP_001128166.2 | 1 - 2 | 656 | 87 | |
GenBank mRNA | BC158625.1 | - | 1 - 2 | 656 | 7454926 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
