Assay Details
| Gene Symbol: LOC691909 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134694.2 | NP_001128166.2 | 7 - 8 | 1164 | 91 | |
| GenBank mRNA | BC158625.1 | - | 7 - 8 | 1164 | 7454926 | 91 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC691909
Genomic Map