Assay Details
| Gene Symbol: Anapc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170435.1 | NP_001163906.1 | 2 - 3 | 97 | 79 | |
| XM_006229860.2 | XP_006229922.1 | 2 - 3 | 330 | 79 | ||
| XM_006229861.2 | XP_006229923.1 | 2 - 3 | 100 | 79 | ||
| XM_006229862.2 | XP_006229924.1 | 2 - 3 | 343 | 79 | ||
| GenBank mRNA | FM103187.1 | - | 2 - 3 | 97 | 79 | |
| FM137052.1 | - | 1 - 2 | 100 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001170435.1, XM_006229860.2, XM_006229861.2, XM_006229862.2 transcripts.
Set Membership: |
|

Anapc15
Genomic Map