Assay Details
| Gene Symbol: Spcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191601.1 | NP_001178530.1 | 2 - 3 | 204 | 98 | |
| GenBank mRNA | FQ214686.1 | - | 2 - 3 | 206 | 98 | |
| FQ227132.1 | - | 2 - 3 | 199 | 98 | ||
| FQ233491.1 | - | 2 - 3 | 202 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spcs2
Genomic Map