Assay Details
| Gene Symbol: Spcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191601.1 | NP_001178530.1 | 4 - 5 | 493 | 120 | |
| GenBank mRNA | FQ214686.1 | - | 4 - 5 | 495 | 120 | |
| FQ232393.1 | - | 1 - 2 | 86 | 120 | ||
| FQ233613.1 | - | 1 - 2 | 86 | 120 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Spcs2
Genomic Map