Assay Details
| Gene Symbol: Cradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108085.1 | NP_001101555.1 | 1 - 2 | 151 | 95 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108085.1 transcripts.
Set Membership: |
|

Cradd
Genomic Map