Assay Details
| Gene Symbol: Glipr1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729783.4 | XP_002729829.3 | 2 - 3 | 826 | 85 | |
| GenBank mRNA | BC083894.1 | - | 2 - 3 | 827 | 85 | |
More Information
Set Membership: |
|

Glipr1l2
Genomic Map