Assay Details
| Gene Symbol: Vezt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006984.1 | NP_001006985.1 | 11 - 12 | 1900 | 113 | |
| XM_006241252.2 | XP_006241314.1 | 11 - 12 | 1947 | 113 | ||
| GenBank mRNA | BC083840.1 | - | 11 - 12 | 1900 | 7114280 | 113 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P133479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153171 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80101 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P133479 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P133479 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P133479 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P138224 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65149 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70238 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 858 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83404 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8483 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52545 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17024 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10901942 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901950 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901945 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901941 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901943 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901949 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901953 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901947 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901944 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901952 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901951 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901946 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901948 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10901940 | - | - |
| Rat Genome 230 2.0 Array | 1397600_at | - | - |
| Rat Genome 230 2.0 Array | 1374589_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374589_at | - | - |
| Rat Genome 230 2.0 Array | 1397600_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1397600_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374589_at | - | - |
| Affymetrix HT Rat Focus Array | 1397600_at | - | - |
| Affymetrix HT Rat Focus Array | 1374589_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700020022.rn.1 | TC0700002511.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700020021.rn.1 | TC0700002511.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700020023.rn.1 | TC0700002511.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374589_at | - | - |
| Rat Expression Set 230 | 1374589_PM_at | - | - |
| Rat Expression Set 230 | 1397600_PM_at | - | - |
| Rat Expression Set 230 | 1397600_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10901940 | - | - |
| Rat Genome U34 Set | rc_AI235367_at | - | - |
| Rat Genome U34 Set | rc_AI170930_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22032226 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359210 | - | - |
| RatRef-12 Expression BeadChip | ILMN_49915 | - | - |

Vezt
Genomic Map