Assay Details
| Gene Symbol: Ears2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760356.1 | XP_008758578.1 | 1 | 466 | 111 | |
| Gene Symbol: Ears2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159493.1 | NP_001152965.1 | 3 - 4 | 533 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ears2l1
Genomic Map