Assay Details
| Gene Symbol: Stx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012700.2 | NP_036832.1 | 1 - 2 | 79 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_012700.2 transcripts.
Set Membership: |
|

Stx1b
Genomic Map